Learn More
Techne™ Stuart™ Skirted 96-well PCR Plates
Description
- Fully skirted to reduce distortion and promote stability
- Available in Black, Clear and White
Specifications
Specifications
| Format | Standard |
| Array | 8 x 12 |
| Color | Black |
| No. of Wells | 96 |
| No. per Pack | 25 |
| Profile | Low |
| Quantity | 25 Pk. |