Learn More
Cole-Parmer™ Stuart™ Shaking Water Bath Stainless Steel Rack
Product Code. 12718988
Change view
Click to view available options
Holds:
120 x 13 mm Culture Tubes
143 x 1.5 mL Microtubes
25 x 50 mL Centrifuge Tubes
30 x 26 mm Culture Tubes
56 x 15 mL Centrifuge Tubes
72 x 16 mm Culture Tubes
Unit Size:
Each
Product Code. Holds unitSize
12718988 56 x 15 mL Centrifuge Tubes Each
10725004 120 x 13 mm Culture Tubes Each
12738988 25 x 50 mL Centrifuge Tubes Each
10181133 143 x 1.5 mL Microtubes Each
12728988 30 x 26 mm Culture Tubes Each
10600394 72 x 16 mm Culture Tubes Each
6 options
Product Code. 12718988
Brand: Cole-Parmer™ 1215297
Only null left
Add to basket
Add to basket
Enhance the versiltility of your Stuart™ water bath
  • Choose from these accessories to make your unit more versatile
  • Designed to be easily interchanged with the stuart water baths and shaking water baths
Specifications
For Use With (Equipment) Shaking Water Bath
Holds 56 x 15 mL Centrifuge Tubes
Material Stainless Steel