Learn More
Cole-Parmer™ Stuart™ Shaking Water Bath Stainless Steel Rack
Artikelnummer. 10600394
Ansicht ändern
Click to view available options
Holds:
120 x 13 mm Culture Tubes
143 x 1.5 mL Microtubes
25 x 50 mL Centrifuge Tubes
30 x 26 mm Culture Tubes
56 x 15 mL Centrifuge Tubes
72 x 16 mm Culture Tubes
Packungsgröße:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Holds Packungsgröße
10600394 72 x 16 mm Culture Tubes Each
10725004 120 x 13 mm Culture Tubes Each
12738988 25 x 50 mL Centrifuge Tubes Each
10181133 143 x 1.5 mL Microtubes Each
12728988 30 x 26 mm Culture Tubes Each
12718988 56 x 15 mL Centrifuge Tubes Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 10600394 Lieferant Cole-Parmer™ Lieferanten-Nr. 1215296
  • ONLINE EXCLUSIVE PRICE
    2980.00 NOK
    2623.54 NOK / Pack of 1
    Save 356.46 NOK
    11% Off
Estimated Shipment: 12-01-2027
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Enhance the versiltility of your Stuart™ water bath
  • Choose from these accessories to make your unit more versatile
  • Designed to be easily interchanged with the stuart water baths and shaking water baths
Spezifikation
For Use With (Equipment) Shaking Water Bath
Holds 72 x 16 mm Culture Tubes
Material Stainless Steel