Learn More
Cole-Parmer™ Stuart™ Heater Block Inserts
Click to view available options
Capacity (Metric):
50 mL
100 mL
250 mL
500 mL
Size:
Large Base
Medium Base
Small and Medium Bases
Unit Size:
Each
Description
- Safest way to heat round-bottom flasks
- Flexible modular system for a wide range of sizes up to 500 mL
- Low-well design is perfect for multi-neck flasks
Specifications
Specifications
| Capacity (Metric) | 250 mL |
| Material | Anodized Aluminum |
| Type | Block |
| Includes | base SHB250BASE |
| Quantity | 1/Ea. |
| Size | Large Base |
| Tube Size | Large |