Learn More
Techne™ Half-skirted 96-well PCR Plate
Product Code. 13068523
Change view
Click to view available options
Color:
Clear
Unit Size:
Pack of 25
Product Code. Color unitSize
13068523 Clear Pack of 25
1 options
Product Code. 13068523
Brand: Techne™ 1621032
Only null left
Add to basket
Add to basket
Semi-skirted PCR plates can be easier to insert and extract than fully skirted plates, and are ideal for use in automated handling or robotic instruments.
  • Semi-skirted
  • Low profile
  • Clear
  • Ideal for automation or robotic use
  • 96-wells
Specifications
Format Standard
Array 8 x 12
Color Clear
Product Type 96-well PCR Plate
Skirt Style Half
No. of Wells 96
No. per Pack 25
Profile Low
Quantity 25 Pk.