Learn More
Cole-Parmer™ Accessories for Cole-Parmer™ MP-100, MP-200, MP-400, and MP-800 Series Melting Point Apparatuses
Accessories for your Cole-Parmer™ MP-100, MP-200, MP-400, or MP-800 Series Melting Point Apparatus.
256.00 NOK - 8645.00 NOK
Specifications
For Use With (Equipment) | SMP10 and SMP20 Digital Melting Point Apparatus |
---|---|
Packaging Quantity | 100 Pk. |
Product Code | Brand | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Price | Quantity & Availability | |||||
12078523
|
Cole-Parmer™
03013-65 |
Closed Glass Melting Tubes |
462.00 NOK
Pack of 100 |
Estimated Shipment: 16-07-2024
Log in to see stock. |
|||||
12058523
|
Cole-Parmer™
03013-64 |
Glass Melting Tubes, One End Open |
268.00 NOK
Pack of 100 |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||||
12048523
|
Cole-Parmer™
03013-63 |
Open-ended Glass Melting Tubes |
256.00 NOK
Pack of 100 |
In Stock
Log in to see stock. |
|||||
11933739
|
Cole-Parmer™
03012-99 |
External Printer |
8645.00 NOK
Each |
Estimated Shipment: 16-07-2024
Log in to see stock. |
|||||
12068523
|
Cole-Parmer™
03013-62 |
Replacement Thermometer |
600.00 NOK
Each |
In Stock
Log in to see stock. |
|||||
Description
- Glass Tubes.
- For MP-100, MP-200, MP-400, and MP-800 Digital Melting Point Apparatus.
- Ideal for educational use.
- Available in packs of 100.
Specifications
SMP10 and SMP20 Digital Melting Point Apparatus | |
100 Pk. |