Learn More
Cole-Parmer™ Tube Holder Disc for Cole-Parmer™ TR-200 Series Tube Rotators
Product Code. 11406558
Change view
Click to view available options
For Use With (Equipment):
For 12 Tubes
For 20 Glaser
For 30 Tubes, 26 mm dia.
For 40 Microcentrifuge Tubes
For 63 Tubes, 12 mm dia.
For 63 Tubes, 16 mm dia.
Unit Size:
Each
Product Code. For Use With (Equipment) unitSize
11406558 For 40 Microcentrifuge Tubes Each
12027144 For 12 Tubes Each
11416558 For 20 Glaser Each
11446558 For 30 Tubes, 26 mm dia. Each
12037144 For 63 Tubes, 12 mm dia. Each
11436558 For 63 Tubes, 16 mm dia. Each
6 options
Product Code. 11406558
Brand: Cole-Parmer™ 0007711
Only null left
Add to basket
Add to basket
Quickly and easily replace tube holder without tools using these Tube Holder Discs for Cole-Parmer™ TR-200 Series Tube Rotators.
  • Choose the tube holder(s) that accommodates your tube size(s).
Specifications
Holds Microcentrifuge tubes
Tube Capacity 1.5 mL
Product Type Tube Holder
For Use With (Equipment) For 40 Microcentrifuge Tubes
No. of Positions 46