Learn More
Cole-Parmer™ Stuart™ Concentrator Evaporator
Click to view available options
Type:
Sample Concentrator
Unit Size:
Each
Description
- Fast efficient sample concentration
- Suitable for all tube configurations
- Compatible with Stuart three block heaters only
- Optional PTFE coating is available for corrosive solutions
Specifications
Specifications
| Type | Sample Concentrator |
| For Use With (Equipment) | SBH130D/3 and SBH200D/3 |