Learn More
Cole-Parmer™ Stuart™ Colony Counter
Product Code. 11983044
Click to view available options
Quantity:
1Ea
Unit Size:
Each
Product Code. 11983044
Brand: Cole-Parmer™ SC6PLUS
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Includes
2 Wolffhuegel graticules and dish centering adapters. The SC6/1/3 Magnifier (seen in the photo) is available as an optional accessory.
Registers cumulative count on the digital display with touch pressure from felt tip marker on petri dish, confirmed by audible tone (can be turned on or off). Stuart™ Colony Counter calculates average count over multiple plates.
Touch pressure with felt tip marker on petri dish registers cumulative count on the digital display with confirmation by audible tone (can be turned on or off). The pressure required to register a count can be adjusted to suit each user. Averaging facility calculates average count over multiple plates. Counting results as well as useful statistics including SD can be sent directly to the accessory printer or to a computer via a USB cable supplied. Sub-stage illumination by low energy bright LED's allows glare-free optimum viewing. A switchable black background is provided to enhance viewing of translucent and difficult to see colonies. Supplied with two Wolffhuegel graticules and dish centering adapters to facilitate use with 50mm to 90mm dishes. A choice of magnifiers and a printer are available as optional accessories.
  • Pressure sensitive counting
  • Average count facility
  • Bright white energy saving LED lighting
  • With BioCote antimicrobial protection
  • Audible count confirmation
  • Choice of light or dark background
  • Connectivity to printer or computer
Specifications
Product Type Colony Counter
Display Type LED
Quantity 1Ea
Total Count 0 to 999 count
Depth (Metric) 300mm
Height (Metric) 140mm
Width (Metric) 310mm
Weight (Metric) 1.5kg
Frequency 50/60Hz
Voltage 90 to 240V
Wattage 70W
For Use With (Application) Cell Counting
Includes 2 Wolffhuegel graticules and dish centering adapters. SC6/1/3 Magnifier (in photo) can be added as an optional accessory.