Learn More
Cole-Parmer™ Stuart™ Borosilicate Glass Reservoir
Click to view available options
:
Capacity: 1400mL
Specifications
Specifications
| Capacity (Metric) | 1400 mL |
| Material | Borosilicate Glass |
| Type | Reservoir |
| Includes | Side filling port chamber |