Learn More
Cole-Parmer™ Stuart™ Borosilicate Glass Receiving Flask
Product Code. 12079121
Change view
Click to view available options
Volume (Metric):
500 mL
1000 mL
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
12079121 1000 mL Each
12089121 500 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12079121 Supplier Cole-Parmer™ Supplier No. 1009668
  • ONLINE EXCLUSIVE PRICE
    4845.00 NOK
    4456.93 NOK / Pack of 1
    Save 388.07 NOK
    8% Off
Estimated Shipment: 23-12-2026
Specifications
Material Borosilicate Glass
Type Receiving Flask
For Use With (Equipment) For RE100 Rotary Evaporator
Quantity 1 Each
Shape Round
Volume (Metric) 1000 mL