Learn More
Cole-Parmer™ Stuart™ Block Heater Insert
Click to view available options
96 well plate
Description
- For use with Stuart block heaters
- Manufactured from anodized aluminum
- Separate hole accommodates a thermometer if desired
Specifications
Description | Holds 96-well plate |
Holds | 96-well plate |
Material | Anodized Aluminum |
No. of Places | 96 |
Type | Block |