Learn More
Strontium Solution for ICP, 1g/L, HNO3 Matrix, Fisher Chemical™
BIOPHARMA
Product Code. 12966224 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
250 mL
500 mL
Packaging:
Bottle
Unit Size:
250mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
12966224 250 mL Bottle 250mL
12976224 500 mL Bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12966224 Supplier Fisher Chemical Supplier No. S 156 250
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Specifications
Packaging Bottle
Analyte or Component Names Sr
Concentration 1±0.002 g/L
Type Mass Spectrometry Standard
Element Sr
Solvent or Matrix Nitric Acid
Quantity 250 mL