missing translation for 'onlineSavingsMsg'
Learn More

Strontium Nitrate, Supelco™

Product Code. 15887620
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15887620 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15887620 Supplier Supelco Supplier No. 1078720250
 Restricted Item
Estimated Shipment: 07-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For analysis EMSURE™

TRUSTED_SUSTAINABILITY

Specifications

Boiling Point 645°C/1013 hPa
Melting Point 570°C
CAS 10042-76-9
Quantity 250 g
Molecular Formula Sr(NO3)2
MDL Number MFCD00011248
Synonym Strontium nitrate, Strontium nitrate
Molecular Weight (g/mol) 211.63
Grade Research
Density 2.986 g/cm3 at 20°C
Chemical Name or Material Strontium Nitrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.