Learn More
Strontium Chloride Hexahydrate, 99-103%, (ACS Reagent Grade), MP Biomedicals
Product Code. 11497892 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11497892 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11497892 Supplier MP Biomedicals Supplier No. 0215258390
  • ONLINE EXCLUSIVE PRICE
    1485.00 NOK
    1411.90 NOK / 500g
    Save 73.10 NOK
    4% Off
 Restricted Item
Estimated Shipment: 07-12-2026
Chemical Identifiers
CAS 10025-70-4
Molecular Formula Cl2H12O6Sr
Molecular Weight (g/mol) 266.61
MDL Number MFCD00149865
InChI Key AMGRXJSJSONEEG-UHFFFAOYSA-L
IUPAC Name strontium(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Sr++]
Specifications
Quantity 500 g
Assay Percent Range 99-103%
Formula Weight 266.6
Grade ACS Reagent
Chemical Name or Material Strontium Chloride Hexahydrate