Learn More
Strontium acetate hemihydrate, 98%
Product Code. 11443207
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
Product Code. 11443207
Brand: Thermo Scientific Alfa Aesar A16149.0E
Request Bulk or Custom Format
CAS: 14692-29-6 | Sr(OOCCH3)2·0.5H2O
PST precursor solutions were prepared from lead (II) acetate,titanium tetraisopropoxide, and strontium acetate hemihydrate with solvents of acetic acid and 2-methoxyethanol.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
PST precursor solutions were prepared from lead (II) acetate,titanium tetraisopropoxide, and strontium acetate hemihydrate with solvents of acetic acid and 2-methoxyethanol.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 14692-29-6
Molecular Formula Sr(OOCCH3)2·0.5H2O
MDL Number MFCD00198064
Synonym Acetic acid strontium salt
Specifications
Melting Point 150°C (in -0.5H2O)
Quantity 2500 g
Merck Index 14,8836
Solubility Information Soluble in water.
Formula Weight 214.72 (205.71 Anhydrous)
Percent Purity 98%
Chemical Name or Material Strontium acetate hemihydrate