Learn More
Cytiva Strip Holder
Product Code. 12598425 Shop All Cytiva Products
Change view
Click to view available options
Size:
7cm
13cm
18cm
24cm
Quantity:
1 Each
6 Pack
Unit Size:
Each
Pack of 6
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Quantity Unit Size
12598425 13cm 6 Pack Pack of 6
10793647 13cm 1 Each Each
10167484 18cm 1 Each Each
10609075 18cm 6 Pack Pack of 6
10782867 24cm 1 Each Each
10036044 7cm 6 Pack Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 12598425 Supplier Cytiva Supplier No. 80641649
Estimated Shipment: 14-01-2027
Specifications
For Use With (Equipment) Ettan IPGphor 3 IEF System
Type Strip Holder
Includes 6 x Strip holder cover, 1 x 60mL strip holder cleaning solution, instructions
Quantity 6 Pack
Size 13cm