Learn More
Ansell™ String Knits™ 76-202 Series White Nylon Light Weight Gloves
Product Code. 12656981 Shop All Ansell Products
Change view
Click to view available options
Size:
6
7
8
9
10
11
Unit Size:
12 pairs
Product Code. Size unitSize
12656981 7 12 pairs
12636981 11 12 pairs
12666981 8 12 pairs
12626981 10 12 pairs
12676981 9 12 pairs
12646981 6 12 pairs
6 options
Product Code. 12656981
Brand: Ansell™ 76202070
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Can be worn on their own or as liners. Permits a longer, more comfortable wear with no weak points which might split or fray.
  • For general handling and delicate manipulation
  • Can also be worn as liners
  • Without seams to rub or irritate
  • Ambidextrous
  • Washable at 40°C
  • Does not shrink, stiffen or lose shape
  • Nylon liner
  • Knitwrist cuff
TRUSTED_SUSTAINABILITY
Specifications
Back Material Nylon
Construction Seamless Knit
Cuff Material Nylon
Product Type Light Weight Gloves
Size 7
Style 76-202
Glove Cuff Color White
Glove Cuff Style Knit
Lining Nylon
Quantity 12 Pairs
Resistant To Mechanical protection
Series 76-202
Voltage Mechanical protection