Learn More
Streptozotocin, TRC
Product Code. 30133813
Change view
Click to view available options
Quantity:
10 mg
50 mg
250 mg
Unit Size:
10mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133813 50 mg 50mg
30133778 250 mg 250mg
30008938 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30133813 Supplier Toronto Research Chemicals Supplier No. TRCS68755050MG
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 18883-66-4
Molecular Formula C8 H15 N3 O7
Molecular Weight (g/mol) 265.22
Synonym 2-Deoxy-2-[[(methylnitrosoamino)carbonyl]amino]-D-glucose, Glucopyranose, 2-deoxy-2-(3-methyl-3-nitrosoureido)-, D- (8CI), Estreptozocin, NSC 37917, NSC 85998, STRZ, STZ, Streptozocin, Streptozoticin, Streptozotocin, U 9889, Zanosar
IUPAC Name 1-methyl-1-nitroso-3-[(3R,4R,5S,6R)-2,4,5-trihydroxy-6-(hydroxymethyl)oxan-3-yl]urea
SMILES CN(N=O)C(=O)N[C@H]1C(O)O[C@H](CO)[C@@H](O)[C@@H]1O
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C8H15N3O7/c1-11(10-17)8(16)9-4-6(14)5(13)3(2-12)18-7(4)15/h3-7,12-15H,2H2,1H3,(H,9,16)/t3-,4-,5-,6-,7?/m1/s1
Formula Weight 265.091
Chemical Name or Material Streptozotocin