missing translation for 'onlineSavingsMsg'
Learn More

Streptozotocin, MP Biomedicals™

Product Code. 11427640 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11427640 500 mg 500mg
11407640 50 mg 50mg
11417640 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11427640 Supplier MP Biomedicals Supplier No. 02100557.5
  • ONLINE EXCLUSIVE PRICE
    2735.00 NOK
    2600.88 NOK / 500mg
    Save 134.12 NOK
    4% Off
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Used in the treatment of pancreatic islet cell tumors. Streptozotocin, MP Biomedicals is a glucosamine-nitrosourea, a DNA-alkylating agent that enters cells via the GLUT2 glucose transport protein. It is used to treat malignant insulinoma and to induce Ty

Streptozocin is used to treat pancreatic islet cell tumors, including malignant insulinoma. It is also used to induce Type 1 diabetes in animals for medical research purposes.

  • An N-nitroso-containing compound that acts as a nitric oxide donor in pancreatic islets
  • Induces death of insulin-secreting cells, producing an animal model of diabetes
  • Potent DNA methylating agent that induces chromosomal breakage
  • Cytotoxic to neuroendocrine tumor cell lines that express the GLUT2 glucose transporter

Chemical Identifiers

CAS 18883-66-4
Molecular Formula C8H15N3O7
Molecular Weight (g/mol) 265.222
InChI Key ZSJLQEPLLKMAKR-UHEPRFFZSA-N
Synonym N-(Methylnitrosocarbamoyl)-α-D-glucosamine, Streptozocin, STZ, 2-Deoxy-2-(3-methyl-3-nitrosoureido)-D-glucopyranose
PubChem CID 45357367
IUPAC Name 1-methyl-1-nitroso-3-[(2R,4R,5S,6R)-2,4,5-trihydroxy-6-(hydroxymethyl)oxan-3-yl]urea
SMILES CN(C(=O)NC1C(C(C(OC1O)CO)O)O)N=O

Specifications

Physical Form Powder
Melting Point 121°C (decomposition) (literature)
Extinction Coefficient EmM (228nm): 6.36 (ethanol)
Quantity 500 mg
Solubility Information Soluble in water (50mg/mL, clear to slightly hazy, light yellow solution) and ethanol.
Formula Weight 265.22
Percent Purity ∼75% α-anomer basis, ≥97%
compliance-icons compliance-icons
Product Identifier
  • Streptozotocin
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 1A
  • Germ cell mutagenicity Category 1A
  • Reproductive toxicity Category 1A
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H340-May cause genetic defects.
  • H350-May cause cancer.
  • H360-May damage fertility or the unborn child.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Streptozotocin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.