Learn More
Streptozotocin, 97+%, Thermo Scientific™
Streptozotocin (streptozocin, CAS # 18883-66-4) is an N-nitrosourea antibiotic produced by Streptomyces achromogenes.
Brand: Thermo Scientific Alfa Aesar J61601.03
| Quantity | 1g |
|---|
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
General Description
• Streptozotocin acts as a bactericidal by alkylating DNA and inhibiting DNA synthesis
• Streptozotocin enters cells through glucose transporter 2
Application
• Streptozotocin induces diabetes in experimental animals and can cause acute liver injury
• It is also useful for research on neuroendocrine tumors and carcinoma cells
Chemical Identifiers
| 18883-66-4 | |
| 265.222 | |
| ZSJLQEPLLKMAKR-UHEPRFFZSA-N | |
| 45357367 | |
| CN(C(=O)NC1C(C(C(OC1O)CO)O)O)N=O |
| C8H15N3O7 | |
| MFCD00006607 | |
| NSC-85998; U-9889 | |
| 1-methyl-1-nitroso-3-[(2R,4R,5S,6R)-2,4,5-trihydroxy-6-(hydroxymethyl)oxan-3-yl]urea |
Specifications
| 18883-66-4 | |
| 1g | |
| C8H15N3O7 | |
| 2060675 | |
| NSC-85998; U-9889 | |
| ZSJLQEPLLKMAKR-UHEPRFFZSA-N | |
| 1-methyl-1-nitroso-3-[(2R,4R,5S,6R)-2,4,5-trihydroxy-6-(hydroxymethyl)oxan-3-yl]urea | |
| 45357367 | |
| ≥97% |
| White | |
| Hygroscopic | |
| MFCD00006607 | |
| 14,8832 | |
| Soluble in water or 100% ethanol | |
| CN(C(=O)NC1C(C(C(OC1O)CO)O)O)N=O | |
| 265.222 | |
| 265.22 | |
| Streptozotocin |
Safety and Handling
- Streptozotocin
Signal Word
- Warning
Hazard Category
- Carcinogenicity Category 2
Hazard Statement
- H351-Suspected of causing cancer.
Precautionary Statement
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contain: Streptozocin