Learn More
Streptozotocin, ≥97%, MP Biomedicals™
Product Code. 11447640 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11447640 5 g 5g
11437640 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11447640 Supplier MP Biomedicals Supplier No. 0210055705
  • ONLINE EXCLUSIVE PRICE
    18255.00 NOK
    17388.74 NOK / 5g
    Save 866.26 NOK
    4% Off
 Restricted Item
Estimated Shipment: 02-10-2026
Chemical Identifiers
CAS 18883-66-4
Molecular Formula C8H15N3O7
Molecular Weight (g/mol) 265.222
InChI Key ZSJLQEPLLKMAKR-UHEPRFFZSA-N
Synonym STZ, 2-Deoxy-2-(3-methyl-3-nitrosoureido)-D-glucopyranose
PubChem CID 45357367
IUPAC Name 1-methyl-1-nitroso-3-[(2R,4R,5S,6R)-2,4,5-trihydroxy-6-(hydroxymethyl)oxan-3-yl]urea
SMILES CN(C(=O)NC1C(C(C(OC1O)CO)O)O)N=O
Specifications
Type Streptozotocin
Physical Form Solid
Color White
Quantity 5 g
Assay Percent Range ≥97%
Formula Weight 265.2
Chemical Name or Material Streptozotocin
compliance-icons compliance-icons
Product Identifier
  • Streptozotocin
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 1A
  • Germ cell mutagenicity Category 1A
  • Reproductive toxicity Category 1A
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H340-May cause genetic defects.
  • H350-May cause cancer.
  • H360-May damage fertility or the unborn child.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Streptozotocin