Learn More
Streptozocin, MedChemExpress
Product Code. 16496306
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16496306 100 mg 100mg
16401706 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16496306 Supplier Medchem Express Supplier No. HY13753
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
Add to basket
MedChemExpress Streptozocin is a potent DNA-methylating antibiotic. Streptozotocin causes methylation of liver and kidney and pancreatic DNA, but no methylation in brain DNA.
Streptozocin is a potent DNA-methylating antibiotic. Streptozotocin causes methylation of liver and kidney and pancreatic DNA, but no methylation in brain DNA.
Specifications
Chemical Name or Material Streptozocin
Physical Form Powder
CAS 18883-66-4
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C8H15N3O7
For Use With (Application) Cancer-programmed cell death
Synonym Streptozotocin U 9889
Solubility Information H2O : 113.3 mg/mL (427.19 mM; Need ultrasonic and warming) ∣DMSO : 83.33 mg/mL (314.19 mM; Need ultrasonic)
SMILES O[C@@H]1[C@@H](CO)O[C@H](O)[C@H](NC(N(C)N=O)=O)[C@H]1O
Molecular Weight (g/mol) 265.22
Formula Weight 265.22
Percent Purity 95.0%
Grade Research