missing translation for 'onlineSavingsMsg'
Learn More

Streptomycin sulfate, Cell Culture Reagent

Product Code. p-7042084
Click to view available options
Quantity:
50 g
100 g
Unit Size:
100g
50g
This item is not returnable. View return policy

Product Code. 15423919

Brand: Thermo Scientific Alfa Aesar J61299.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Streptomycin sulfate, CAS # 3810-74-0, is the sulfate salt form of streptomycin, and an aminoglycoside with antibacterial activity. It also shows a protein synthesis inhibitory activity. | CAS: 3810-74-0 | C42H84N14O36S3 | 1457.376 g/mol

Inhibits initiation and causes misreading of rRNA in protein synthesis Streptomycin sulfate is used to study the mechanisms of streptomycin resistance. It is used to inhibit bacterial contamination in cell culture applications in association with penicillin. It is an antibiotic drug initially used for tuberculosis treatment. It inhibits protein synthesis by binding to 30S ribosomal subunit, which induces the misreading of the genetic code. It is involved in the initiation of translation of DNA, thereby results in death for a susceptible bacterium. Further, it controls bacterial and fungal diseases of certain fruit, vegetables, seed, ornamental crops and algae in ornamental ponds and aquaria

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Streptomycin sulfate is an aminoglycoside sulfate salt derived from Streptomyces griseus that shows antibacterial activity.
  • It inhibits the initiation and causes misreading of rRNA in protein synthesis. This compound binds to the S12 protein of the bacterial 30S ribosomal subunit, thereby inhibiting peptide elongation and protein synthesis.

Application

  • Streptomycin sulfate can be used to study the mechanisms of streptomycin resistance
  • This compound can be used in laboratory applications combined with penicillin to inhibit bacterial contamination in in vitro cell culture
  • Streptomycin sulfate shows antibiotic activity against Mycobacterium tuberculosis
  • This compound can be used as a protein synthesis inhibitor
  • It acts against gram-negative and gram-positive bacteria
  • Streptomycin sulfate also shows a fungicide activity
  • It can be used to remove nucleic acids from a biological extract
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3810-74-0
Molecular Formula C42H84N14O36S3
Molecular Weight (g/mol) 1457.376
MDL Number MFCD00037023
InChI Key QTENRWWVYAAPBI-FFCQDDOVSA-N
PubChem CID 124080941
IUPAC Name 2-[(1R,2S,3S,4S,5R,6S)-3-(diaminomethylideneamino)-4-[(2R,3S,4S,5S)-3-[(2R,3S,4R,5R,6S)-4,5-dihydroxy-6-(hydroxymethyl)-3-(methylamino)oxan-2-yl]oxy-4-formyl-4-hydroxy-5-methyloxolan-2-yl]oxy-2,5,6-trihydroxycyclohexyl]guanidine;2-[(1R,2S,3S,4R,5R,6R)-3-(
SMILES CC1C(C(C(O1)OC2C(C(C(C(C2O)O)N=C(N)N)O)N=C(N)N)OC3C(C(C(C(O3)CO)O)O)NC)(C=O)O.CC1C(C(C(O1)OC2C(C(C(C(C2O)O)N=C(N)N)O)N=C(N)N)OC3C(C(C(C(O3)CO)O)O)NC)(C=O)O.OS(=O)(=O)O.OS(=O)(=O)O.OS(=O)(=O)O

Especificaciones

Type Streptomycin Sulfate
Physical Form Powder
Color White
Quantity 50 g
Beilstein 3894995
Merck Index 14,8826
Solubility Information Soluble in water.
Formula Weight 728.69
Grade Cell Culture
Chemical Name or Material Streptomycin sulfate
compliance-icons compliance-icons
Nombre del producto
  • Streptomycin sulfate, Cell Culture Reagent
Palabra de advertencia
  • Warning
Clasificación
  • Acute toxicity Category 4
  • Reproductive toxicity Category 2
  • Skin sensitiser Category 1
Indicaciones de peligro
  • H302-Harmful if swallowed.
  • H317-May cause an allergic skin reaction.
  • H361d-Suspected of damaging the unborn child.
Consejos de prudencia
  • P201-Obtain special instructions before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Otros peligros
  • MIXTURE LIST-Contains : Streptomycin sulfate

RUO – Research Use Only

Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado