Learn More
IBA Lifesciences StrepMAB-Immo
Product Code. 12676817 Shop All IBA Lifesciences Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12676817 100 μg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12676817 Supplier IBA Lifesciences Supplier No. 21517001
  • ONLINE EXCLUSIVE PRICE
    5485.00 NOK
    4384.47 NOK / Pack of 1
    Save 1100.53 NOK
    20% Off
 Restricted Item
Estimated Shipment: 14-10-2026
StrepMAB-Immo is a monoclonal antibody to detect Strep-tag™II and Twin-Strep-tag™ fusion proteins
StrepMAB-Immo is a murine, high-affinity monoclonal antibody, which is especially suited for stable, mild, and oriented capturing of Strep-tag™II and Twin-Strep-tag™ fusion proteins on solid phases (e.g. Biacore CM5 sensor chips) or for detection of those proteins in applications like ELISA, FACS, immunohistochemistry, and immunocytochemistry. The nearly irreversible binding is validated for fusion proteins carrying a C- or N-terminal Strep-tag™II providing an N-terminal extension by a SerAla linker (recombinant protein-SA-WSHPQFEK or SA-WSHPQFEK-recombinant protein). Proteins containing the Strep-tag™II with N-terminal sequences other than SerAla are bound with reduced affinity in the cases tested so far, the Twin-Strep-tag™ always contains the SerAla linker (WSHPQFEK-GGGS-GGGS-GGSA-WSHPQFEK). Please note: For detection of Strep-tag™II or Twin-Strep-tag™ fusion proteins in western blots, the use of StrepMAB-Classic or Strep-Tactin™ is recommended.
Specifications
Item Description Lyophilized powder
Product Type Monoclonal Antibody
For Use With (Application) ELISA, FACS, Immunofluorescence, SPR
Storage Requirements -20°C
Form Suspension in PBS
Quantity 100 μg
For Research Use Only.