Learn More
IBA Lifesciences GmbH Strep-Tactin™ PE for Fab Streptamers™
Shop All IBA Lifesciences Products
Click to view available options
:
Includes: All required reagents for cell isolation
Description
- Low-affinity Fab-Strep molecules are reversibly multimerized on Strep-Tactin™ PE or APC forming a fluorescent Fab Streptamer™ for cell staining
- Treatment of labeled target cells with the competing Strep-Tactin™ ligand biotin causes a disruption of the Fab Streptamer™ complex and results in spontaneous dissociation of the Fab-Streps from the target cells
- The result is a functional and label-free target cell
Specifications
Specifications
| Includes | All required reagents for cell isolation |
| For Use With (Application) | For 5 x 107 to 1 x 108 cells |