Learn More
Thermo Scientific Chemicals Straight Wall Zirconium Crucible
Click to view available options
Capacity (Metric):
10 mL
15 mL
20 mL
25 mL
35 mL
55 mL
75 mL
100 mL
250 mL
500 mL
1000 mL
Diameter (Metric) Outer:
Outside: 101mm
Outside: 127mm
Outside: 27mm
Outside: 33mm
Outside: 45mm
Outside: 46mm
Outside: 47mm
Outside: 51mm
Outside: 59mm
Outside: 82mm
Height (Metric):
18mm
22mm
29mm
34mm
41mm
45mm
59mm
65mm
89mm
Unit Size:
Each
Descripción
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Especificaciones
Especificaciones
| Description | Straight Wall |
| Material | Zirconium |
| Capacity (Metric) | 75 mL |
| Diameter (Metric) Outer | Outside: 51mm |
| Height (Metric) | 29mm |
| Product Type | Zirconium Crucible |
RUO – Research Use Only
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido