Learn More
Thermo Scientific Chemicals Straight Wall Inconel Crucible;Cap (ml), 45;Outside Dia (mm), 46;Depth (mm), 34
Product Code. 15473635
Change view
Click to view available options
Capacity (Metric):
5 mL
10 mL
15 mL
20 mL
25 mL
55 mL
75 mL
100 mL
250 mL
500 mL
1000 mL
Diameter (Metric) Outer:
Outside: 101mm
Outside: 127mm
Outside: 21mm
Outside: 27mm
Outside: 33mm
Outside: 45mm
Outside: 46mm
Outside: 47mm
Outside: 51mm
Outside: 59mm
Outside: 82mm
Unit Size:
Each
Product Code. Capacity (Metric) Diameter (Metric) Outer unitSize
15473635 500 mL Outside: 101mm Each
15466135 5 mL Outside: 21mm Each
15486135 10 mL Outside: 27mm Each
15406145 15 mL Outside: 33mm Each
15426145 20 mL Outside: 33mm Each
15446145 25 mL Outside: 45mm Each
15476145 55 mL Outside: 46mm Each
15423625 1000 mL Outside: 127mm Each
15496145 100 mL Outside: 59mm Each
15426155 55 mL Outside: 47mm Each
15456155 75 mL Outside: 51mm Each
15461437 250 mL Outside: 82mm Each
12 options
Product Code. 15473635
Brand: Thermo Scientific Chemicals 036090.KT
Request Bulk or Custom Format
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Description Straight Wall
Material Inconel
Capacity (Metric) 500 mL
Diameter (Metric) Outer Outside: 101mm
Product Type Inconel Crucible