Description
- Range of different convenient lengths
Specifications
Specifications
| Tip Style | Straight |
| Length (English) | 7.9 in. |
| Material | Stainless Steel (316L) |
| Length (Metric) | 200 mm |
| Product Type | Tweezers |
| Quantity (No. per Case) | 1 Tweezer/Cs. |
| Sterility | Non-sterile |
| Style | Standard |