Learn More
Sampling Systems™ Straight Tweezers
Product Code. 17912112 Shop All Sampling Systems Products
Change view
Click to view available options
Length (Metric):
100 mm
200 mm
300 mm
400 mm
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
17912112 300 mm Each
17992102 100 mm Each
17942132 200 mm Each
17932132 400 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 17912112 Supplier Sampling Systems™ Supplier No. A596300
Top Sellers. Mega Savings.
Use promo code 27997
View all promotions
Estimated Shipment: 05-01-2027
These tweezers made of 316L stainless steel are designed for use in laboratory, production and industrial environments. Tweezers make it easy to pick up intricate items, such as vials and tissues.
  • Range of different convenient lengths
Specifications
Product Type Tweezers
Length (English) 11.8 in.
Length (Metric) 300 mm
Material Stainless Steel (316L)
Quantity 1 Tweezer/Cs.
Sterility Non-sterile
Style Standard
Tip Style Straight