Learn More
Lenz Laborglasinstrumente™ Straight Tubing Connectors
Product Code. 10469552
Change view
Click to view available options
Diameter (Metric) Outer:
10 mm
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Outer Unit Size
10469552 10 mm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10469552 Supplier Lenz Laborglasinstrumente™ Supplier No. 01600010
  • ONLINE EXCLUSIVE PRICE
    125.00 NOK
    108.92 NOK / Pack of 10
    Save 16.08 NOK
    12% Off
Estimated Shipment: 11-12-2026
Manufactured from DURAN™ Lenz Laborglasinstrumente™ Straight Tubing Connectors are ideal for connecting tubes of various diameters.
  • Tubing connectors
  • Straight
  • With two glass olives
  • Made of DURAN™ tubing
Specifications
Diameter (Metric) Outer 10 mm
Overall Length (Metric) 115 mm
Packaging Quantity 10