Learn More
Lenz Laborglasinstrumente™ Splash Heads Acc. to Stutzer, Straight
Product Code. 11953461
Change view
Click to view available options
Standard Taper Joint:
NS 14/23 NS 14/23
NS 29/32, NS 14/23
NS 29/32, NS 29/32
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Standard Taper Joint Unit Size
11953461 NS 29/32, NS 14/23 Each
10588421 NS 14/23 NS 14/23 Each
10231071 NS 29/32, NS 29/32 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11953461 Supplier Lenz Laborglasinstrumente™ Supplier No. 05035045
  • ONLINE EXCLUSIVE PRICE
    1090.00 NOK
    1030.33 NOK / Pack of 1
    Save 59.67 NOK
    5% Off
Estimated Shipment: 18-12-2026
For placement onto distillation flasks for separation of drops. Lenz Laborglasinstrumente™ Splash Heads Acc. to Stutzer, Straight, are manufactured from durable Duran™ glass tubing.
  • Straight with NS cone and socket
  • Temperature and chemical resistance
  • Minimal thermal expansion
Specifications
For Use With (Application) For separation of drops (Direct placement onto distillation flasks)
Type Straight Splash Heads
Material Duran Glass
Standard Taper Joint NS 29/32, NS 14/23