Learn More
Gosselin™ Straight Specimen Containers with Blue Cap and Label
CORNING GOSSELIN™ Straight Container, Specimen Vials, PP come in different sizes and quantities, and are available sterile and non-sterile.
1711.66 NOK - 2283.06 NOK
Specifications
Product Type | Specimen Container |
---|---|
Closure Included | Yes |
Material | Polypropylene |
Shape | Straight |
Sterility | Sterile |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
15498804
|
Gosselin™
TP30C-021 |
40 mL |
2283.06 NOK
Pack of 1000 |
Estimated Shipment: 19-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15488814
|
Gosselin™
TP35C-021 |
60 mL |
1711.66 NOK
Pack of 700 |
Estimated Shipment: 19-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Clear and break-resistant polypropylene (PP)
- Available in different sizes and quantities
- Color-coded screw caps
Specifications
Specimen Container | |
Polypropylene | |
Sterile | |
Screw Cap | |
White label |
Yes | |
Straight | |
Blue | |
Clear Blue | |
White label |