Learn More
BRAND™ Straight Burette Stopcock with Spare Key
Product Code. 10682052 Shop All Brand Products
Change view
Click to view available options
Capacity (Metric):
50 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10682052 50 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10682052 Supplier BRAND™ Supplier No. 81939
  • ONLINE EXCLUSIVE PRICE
    626.00 NOK
    528.34 NOK / Pack of 1
    Save 97.66 NOK
    15% Off
Estimated Shipment: 07-12-2026
Stopcock housing with precision Boro 3.3 burette tip. Interchangeable PTFE key with retention device.
  • Amber color
  • Straight stopcock for burette cap
  • Includes PTFE key
  • Stopcock size 3NS/12
TRUSTED_SUSTAINABILITY
Specifications
Color Amber
Capacity (Metric) 50 mL
For Use With (Equipment) Burette
Material Borosilicate Glass, PTFE
No. per Pack 1 Each
Includes Burette Cap
Type Stopcock