Learn More
BRAND™ Straight Burette Stopcock with PTFE Key
Product Code. 10461253 Shop All Brand Products
Change view
Click to view available options
Material:
Amber Borosilicate Glass (3.3)
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
10461253 Amber Borosilicate Glass (3.3) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10461253 Supplier BRAND™ Supplier No. 81937
  • ONLINE EXCLUSIVE PRICE
    626.00 NOK
    564.62 NOK / Pack of 1
    Save 61.38 NOK
    9% Off
Estimated Shipment: 21-12-2026
Straight Burette Stopcock for Burette Cap includes an interchangeable PTFE key.
  • Stopcock housing with precision Borosilicate Glass 3.3 burette tip
  • Interchangeable PTFE key with retention device
  • Stopcock size 3NS/12
TRUSTED_SUSTAINABILITY
Specifications
Type Burette Stopcock
Material Amber Borosilicate Glass (3.3)
For Use With (Equipment) 25 mL Burette
Includes PTFE Key