Learn More
Thermo Scientific™ Storage Vials
Product Code. 12458676
Change view
Click to view available options
Color:
Amber
Clear
White
White, Clear
Quantity:
100
100 Caps
100 Septa
100 Vials
200
200 (vial and caps are sold separately)
200 Vials
Unit Size:
Pack of 100
Pack of 200
Volume (Metric):
2 mL
4 mL
8 mL
12 mL
16 mL
22 mL
40 mL
Closure Size:
13-425
15 mm
15-425
18 mm
18-400
20-400
24 mm
24-400
8-425
Product Code. Color Quantity unitSize Volume (Metric) Closure Size
12458676 White, Clear 100 Pack of 100 - -
12488676 Clear 100 Vials Pack of 100 2 mL 8-425
11971583 Clear 100 Pack of 100 4 mL -
11905627 Clear 200 Pack of 200 8 mL -
12005097 Clear 200 Pack of 200 12 mL -
11508250 Clear 200 Pack of 200 16 mL -
12438686 Clear 200 (vial and caps are sold separately) Pack of 200 22 mL -
12201182 Clear 100 Pack of 100 40 mL -
11528250 Amber 100 Pack of 100 4 mL -
13141673 Amber 200 Pack of 200 8 mL -
12498676 Amber 200 Pack of 200 12 mL -
11518250 Amber 200 Pack of 200 16 mL -
12458686 Amber 100 Pack of 100 40 mL -
11995617 White 100 Pack of 100 - 15 mm
11548250 White 100 Pack of 100 16 mL 18 mm
12438636 White 100 Caps Pack of 100 - 20-400
12670356 White 100 Pack of 100 - 24 mm
12478676 White, Clear 100 Pack of 100 - -
12418676 White, Clear 100 Septa Pack of 100 - 13-425
12428676 White, Clear 100 Septa Pack of 100 - 15-425
12438676 White, Clear 100 Septa Pack of 100 - 18-400
12448676 White, Clear 100 Pack of 100 - -
12468676 White, Clear 100 Pack of 100 - -
11593740 Amber 100 Vials Pack of 100 40 mL 24-400
11553740 Amber 200 Vials Pack of 200 16 mL 18-400
11533740 Amber 200 Vials Pack of 200 12 mL 15-425
11523740 Amber 200 Vials Pack of 200 8 mL 15-425
10445082 White 100 Pack of 100 - 24 mm
10001802 White 100 Caps Pack of 100 - 24-400
10462202 White 100 Pack of 100 - 18 mm
10356592 White 100 Pack of 100 - 15 mm
11583740 Clear 100 Vials Pack of 100 40 mL 20-400
11563740 Clear 200 Pack of 200 22 mL -
11543740 Clear 200 Pack of 200 16 mL -
11513740 Clear 200 Pack of 200 8 mL -
10296774 Clear 200 Pack of 200 12 mL -
13121673 Amber 100 Pack of 100 2 mL -
37 options
Product Code. 12458676
Brand: Thermo Scientific™ B799524
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Store your samples in these vials to eliminate leaching of ions while maintaining a consistent pH for the life of your sample. Thermo Scientific™ National™ and Thermo Scientific™ Chromacol™ Sample Storage Screw Thread Vials are manufactured from superior quality 33 expansion borosilicate clear (Type 1, Class A) or 51A amber (Type 1, Class B) glass. Storage vials range from 2mL to 40mL.
  • PTFE-lined solid-top storage caps
  • PTFE on film/foam backing offers broad chemical resistance
  • Not autoclavable
Order as individual sample storage vials and closures or as unassembled convenience kits.
Unassembled convenience kits saves time during sample preparation.
  • Solid-top convenience kits include matched quantities of vials and screw caps with pre-assembled septa.
  • Open-top convenience kits contain shrink-wrapped vials and separately packaged caps and septa in polybags.
TRUSTED_SUSTAINABILITY
Specifications
Description Septa for 24-400 Screw Cap
Color White, Clear
Material PTFE, Silicone
Quantity 100
Thickness (Metric) 2.6 mm
Product Type Septum Only