Learn More
Thermo Scientific™ Storage Canes
Description
Store Thermo Scientific™ Nalgene™ or Thermo Scientifc™ Nunc™ cryostorage tubes in Thermo Scientific™ Storage Canes, available in standard or stepped format to accommodate a variety of vial sizes and designs.
- Able to store tubes protected with Thermo Scientific™ CryoFlex™ tubing
- Compatible with color coders that attach to the end of the cane and can be written on for quick identification and inventory management
Specifications
Specifications
| Design | Coder |
| Color | White |
| Material | Aluminum |
| Type | Cryo Cane Coder |
| Quantity | 100/Pk., 100/Cs. |
| Length (Metric) | Not Applicable |