Learn More
Buerkle™ Storage Bottles without Threaded Connector
Shop All Bürkle Products
Click to view available options
Capacity (Metric):
5 L
10 L
25 L
60 L
Unit Size:
Each
Description
- The venting valve (accessory) lets air into the container in order to prevent a vacuum in the bottle
- The canister can also be used as a settling canister
- HDPE
- Bottle opening diameter 50mm
Specifications
Specifications
| Capacity (Metric) | 60 L |
| Color | Transparent |
| For Use With (Application) | Foodstuffs |
| Includes | Closure, 2 handles |
| Material | HDPE |
| Quantity | 1 |
| Transparency | Yes |
| Thread Size (Metric) | 50 mm |
| Closure Included | Yes |
| Graduated | Yes |
| Show More |