Learn More
ratiolab™ Stoppers for Test Tubes
Product Code. p-200106389
Click to view available options
Color:
Blue
Green
Red
Yellow
Diameter (Metric) Outer:
11 to 12 mm
16 to 17 mm
Unit Size:
Pack of 1000
Product Code. 12341523
Brand: ratiolab™ 391490R
Only null left
Add to basket
Add to basket
Close your test tubes with these highly secure stoppers.
  • Provides very tight seal due to two sealing rings
Specifications
Diameter (Metric) Outer 16 to 17 mm
Material Polyethylene
Color Red
Packaging Bag
Quantity 1000 Stoppers/Bag