Learn More
ratiolab™ Stoppers for Test Tubes
Product Code. 11331153
Change view
Click to view available options
Color:
Blue
Green
Red
Yellow
Diameter (Metric) Outer:
11 to 12 mm
16 to 17 mm
Unit Size:
Pack of 1000
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Diameter (Metric) Outer Unit Size
11331153 Blue 16 to 17 mm Pack of 1000
12341523 Red 16 to 17 mm Pack of 1000
14809860 Yellow 16 to 17 mm Pack of 1000
13465056 Green 11 to 12 mm Pack of 1000
13470147 Green 16 to 17 mm Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 11331153 Supplier ratiolab™ Supplier No. 391490B
  • ONLINE EXCLUSIVE PRICE
    240.00 NOK
    156.83 NOK / Pack of 1000
    Save 83.17 NOK
    34% Off
 Restricted Item
Estimated Shipment: 29-12-2026
Close your test tubes with these highly secure stoppers.
  • Provides very tight seal due to two sealing rings
Specifications
Diameter (Metric) Outer 16 to 17 mm
Material Polyethylene
Color Blue
Packaging Bag
Quantity 1000 Stoppers/Bag