Learn More
Lenz Laborglasintrumente Stoppers Octagonal Plate, Plastic
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 01401010
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 10224531
  • 80.72 NOK / Pack of 10
Estimated Shipment: 23-07-2024
to see stock.
Add to basket
Description
Description
  • Acc. to DIN 12252
  • Form A and E
  • Solid or semi-hollow
  • Material: Plastic
Specifications
Specifications
White, Red
Polyethylene
NS 10/19