Learn More
Thermo Fisher Scientific™ FermionX Stomacher™ 400 Bag Opener
Product Code. 12764876
Change view
Click to view available options
No. of Positions:
1
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Positions Unit Size
12764876 1 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12764876 Supplier Thermo Fisher Scientific™ Supplier No. BA6095
  • ONLINE EXCLUSIVE PRICE
    577.00 NOK
    532.86 NOK / Pack of 1
    Save 44.14 NOK
    7% Off
Estimated Shipment: 09-12-2026
The Stomacher™ 400 bag opener from FermionX holds open bags for easy addition of samples without risking instrument contact on the bag.
  • Compatible with Stomacher™ 80 and Stomacher™ 400 bag sizes
  • Stable on benchtop or scales
  • Easily cleaned with most lab disinfectant
  • Capacity: One place
  • Dimensions (L x W x H): 15.5 x 11.5 x 27 cm
Specifications
Product Type Bag Opener
Material Plastic polymer
For Use With (Equipment) Stomacher™ 80 and Stomacher™ 400 bag
No. of Positions 1