Learn More
Kinematica™ Stodger for Polymix™ MFC 90 D Mill
Product Code. 11940359 Shop All Kinematica Products
Click to view available options
For Use With (Equipment):
PX-MFC 90 D
Unit Size:
Each
Product Code. 11940359
Brand: Kinematica™ 35080001
For active insertion of fibrous materials, which do not reach the grinding chamber due to their weight
Specifications
Product Type Stodger
For Use With (Application) Active insertion of fibrous materials, which do not reach the grinding chamber due to their weight
For Use With (Equipment) PX-MFC 90 D