missing translation for 'onlineSavingsMsg'
Learn More

STK38 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 55335-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16846815

  • 3828.53 NOK / 150µL
Estimated Shipment: 17-10-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This antibody recognizes the C-terminal of STK38.

Stk38,also known as NUCLEAR DBF2-RELATED PROTEIN, contains 1 protein kinase domain that interacts with mob1 and mob2.The homodimeric s100b binds to two molecules of stk38. It is ubiquitously expressed with highest levels observed in peripheral blood leukocytes. Stk38 activated by binding of s100b which releases autoinhibitory n-lobe interactions, enabling atp to bind and the autophosphorylation of ser-281. Thr-444 then undergoes calcium- dependent phosphorylation by an upstream kinase. Interactions between phosphorylated Thr-444 and the n-lobe promote additional structural changes that complete the activation of the kinase. Autoinhibition is also released by the binding of mob1/mobkl1a and mob2/hcca2 to the n-terminal of stk38.
Specifications

Specifications

STK38
Polyclonal
Unconjugated
STK38
NDR, NDR1, NDR1 protein kinase, Nuclear Dbf2 related kinase 1, serine/threonine kinase 38, STK38
Rabbit
Antigen Affinity Chromatography
RUO
106504, 11329
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.71 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q15208, Q91VJ4
Stk38
Peptide
150 μL
Primary
Human, Mouse
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers