missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STK38 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55335-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody recognizes the C-terminal of STK38.
Stk38,also known as NUCLEAR DBF2-RELATED PROTEIN, contains 1 protein kinase domain that interacts with mob1 and mob2.The homodimeric s100b binds to two molecules of stk38. It is ubiquitously expressed with highest levels observed in peripheral blood leukocytes. Stk38 activated by binding of s100b which releases autoinhibitory n-lobe interactions, enabling atp to bind and the autophosphorylation of ser-281. Thr-444 then undergoes calcium- dependent phosphorylation by an upstream kinase. Interactions between phosphorylated Thr-444 and the n-lobe promote additional structural changes that complete the activation of the kinase. Autoinhibition is also released by the binding of mob1/mobkl1a and mob2/hcca2 to the n-terminal of stk38.Specifications
| STK38 | |
| Polyclonal | |
| Unconjugated | |
| STK38 | |
| NDR, NDR1, NDR1 protein kinase, Nuclear Dbf2 related kinase 1, serine/threonine kinase 38, STK38 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 106504, 11329 | |
| -20°C | |
| Liquid |
| Western Blot, Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| 0.71 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q15208, Q91VJ4 | |
| Stk38 | |
| Peptide | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction