Learn More
Cole-Parmer™ Stuart™ Stirring Hotplate and Hotplate Accessories
Product Code. p-7232468
Change view
Click to view available options
For Use With (Equipment):
Undergrad Hot Plates, Stirrers
Type:
Temperature Controller
Unit Size:
Each
Product Code. For Use With (Equipment) Type unitSize
17342681 Undergrad Hot Plates, Stirrers Temperature Controller Each
1 options
Product Code. 17342681
Brand: Cole-Parmer™ Stuart™ 0480790
Only null left
Add to basket
Add to basket
Replacement parts and accessories for use with Hotplates, stirrers, and/or stirring Hotplates.
  • Replacement parts and accessories for various Hotplates, stirrers, and stirring Hotplates
Specifications
Description Plugs into hotplate or stirrer
For Use With (Equipment) Undergrad Hot Plates, Stirrers
For Use With (Application) Accurate liquid temperature control
Length (English) 3 in.
Model SCT2
Temperature (English) 122°F to 392°F
Temperature (Metric) 50°C to 200°C
Type Temperature Controller
Width (English) 3.5 in.
Weight (Metric) 635 g
Display LED
Dimensions (L x W x H) 7.5 x 9 x 12.3 cm
Accuracy ±0.5°C
Coating Uncoated
Material Plastic, Stainless Steel
Weight (English) 1.4 lb.