Learn More
SP Bel-Art™ Stirring Bars Spinbar™
Multi-faceted shape provides greater surface area and added turbulence for more vigorous, thorough mixing when compared to the smooth surface of cylindrical bars
Brand: SP Bel-Art™ F37110-1608
Additional Details : Weight : 0.78810kg
Description
- With pivot ring for better mixing conditions
- Alnico magnet encapsulated with PTFE™ brand PTFE coating
- FDA Grade, USP Class VI
- Good chemical compatibility
- Autoclavable
Specifications
8 mm | |
10/Pk. | |
16 mm | |
Octagon |