Learn More
Lenz Laborglasintrumente Stirrer with Stirrer Bearing
Product Code. 10015404
Change view
Click to view available options
Material:
Stainless Steel
Unit Size:
Each
Product Code. Material unitSize
10015404 Stainless Steel Each
1 options
Product Code. 10015404
Brand: Lenz Laborglasintrumente 06310506
Only null left
Add to basket
Add to basket
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Shaft with blade and PTFE security screw
  • Gastight closure of vacuum stirring apparatuses up to 10-3 mbar at a continuous operation speed of max.
  • 800 rpm, for round bottom flasks 250-2000 mL
  • Shaft made of DURAN™ tubing
Specifications
Description Pressure spring
For Use With (Equipment) For round bottom flasks 250 to 2000 mL
Material Stainless Steel