Learn More
Lenz Laborglasinstrumente™ Stirrer Bearings with a Dynamic Double Seal
Click to view available options
Tubing Size:
Joint (NS) 29/32
Joint (NS) 45/46
Unit Size:
Each
Description
- With extended guidance for reduced vibrations and a longer lifetime
- Made of borosilicate glass
- PTFE(sealings)
Specifications
Specifications
| Diameter (Metric) | 16 mm |
| Material | Borosilicate Glass |
| Tubing Size | Joint (NS) 29/32 |
| Material (Seal) | PTFE |
Product Content Correction
Product Title
Lenz Laborglasinstrumente™ Stirrer Bearings with a Dynamic Double Seal >