Learn More
Cole-Parmer™ Stir Bars
Product Code. 12799038
Change view
Click to view available options
Length (Metric):
10 mm
12 mm
Diameter (Metric):
4.5 mm
6 mm
Shape:
Cylinder
Oval
Unit Size:
Pack of 10
Product Code. Length (Metric) Diameter (Metric) Shape unitSize
12799038 12 mm 4.5 mm Cylinder Pack of 10
12789038 10 mm 6 mm Oval Pack of 10
2 options
Product Code. 12799038
Brand: Cole-Parmer™ 3663099
Only null left
Add to basket
Add to basket
Stir Bars suitable for use with Cole-Parmer™ Integrity 10, RS-200, or RS-250 series reaction stations.
  • Suitable for use with Cole-Parmer™ Integrity 10, RS-200, or RS-250 series reaction stations.
  • Choose from a wide range of accessories suitable for your reaction station including reaction vessels, probes, and software.
Specifications
Diameter (English) 0.18 in.
For Use With (Application) For Stem Integrity
For Use With (Equipment) Integrity 10, RS-200, and RS-250 Reaction Stations
Length (English) 0.5 in.
Diameter (Metric) 4.5 mm
Length (Metric) 12 mm
Shape Cylinder
Type Magnetic Stir Bar